ContactUsLinkComponent
Mikrostik™ Non-Conductive Adhesive, Electron Microscopy Sciences
Mikrostik™ Non-Conductive Adhesive, Electron Microscopy Sciences
Catalog # 103301-908
CAS Number:  
undefined
Mikrostik™ Non-Conductive Adhesive, Electron Microscopy Sciences
Catalog # 103301-908
Supplier Number:  12646-01
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Adhesive, Non – Conductive
  • Size:
    14 mL
  • Cat. no.:
    103301-908
  • Supplier No.:
    12646-01
Specifications
About this item
Fast drying, ultra thin clear adhesive.
  • Suitable for mounting small particles which can be submerged in other adhesives
  • Non-Conductive
  • Can dilute with methyl ethyl ketone
  • Bottle with applicator brush