If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Eyelash Manipulator
- Cat. no.:103302-486
- Supplier No.:71182
Specifications
About this item
This is for direct manipulation of sections and small tissue samples.
- Sable brush handle made from the highest quality wood
- Cleaned and sterilized with alcohol and a tissue
Suitable for ultramicrotomy to carefully tease sections that are in the diamond knife boat.