Tweezers WA1 Handle with 3-STD Tips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
Swiss made precision tweezers from Dumont.
- Hand made of the highest quality steels and alloys
- Perfectly finished to suit your application
These tweezers have an anti-magnetic WA1 handle with replaceable 3-STD tips. Tips attach with screws. Length is 125 mm.
Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as Inox.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together