Swiss made precision tweezers from Dumont.
- Hand made of the highest quality steels and alloys
- Perfectly finished to suit your application
These tweezers have an anti-magnetic WA1 handle with replaceable 2-STD tips. Tips attach with screws. Length is 130 mm.
Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as Inox. Resistant up to temperatures of 400°C (DIN 50 914) and suitable for autoclave sterilization at 270 °C.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.