Order Entry
Canada
ContactUsLinkComponent
Quick-DNA™ Fungal/Bacterial Kits, Zymo Research
Quick-DNA™ Fungal/Bacterial Kits, Zymo Research
Supplier:  Zymo Research
Certificates
undefined
Quick-DNA™ Fungal/Bacterial Kits, Zymo Research
Supplier:  Zymo Research

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Yeast/Fungal DNA
    Bacterial DNA
  • Sample type:
    Bacteria
    Yeast/Fungi
  • Target molecule:
    DNA
  • Description:
    Quici-DNA™ Fungal/Bacterial Midiprep Kit

Specifications

Frequently Bought Together