About this item
Quick recovery of viral DNA from a wide range of sources.
- Quick recovery of viral DNA from a wide range of sources using Zymo-Spin™ Column and Plate technologies
- Column and plate designs allow DNA to be eluted at high concentrations into minimal volumns
- Eluted DNA is suitable for PCR, Southern blotting, and restriction endonuclease digestion
The Quick-DNA™ Viral Kits provide for the rapid isolation of high-quality viral DNA from a wide range of biological sources. A uniquely designed buffer is included for the efficient denaturation of viral particles in whole blood (fresh and stored), plasma, serum, tissue, ascites, cultured cells, and liquid samples. DNA can be eluted with elution buffer or water, and is ideal for subsequent PCR, nucleotide blotting, and restriction endonuclease digestion procedures.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Method / Format:Multiwell plate
- Prep size:Mini
- Refine target molecule:Total DNA
- Sample type:BloodCultured cellsHuman/animal cells and tissues
- Target molecule:DNA
- Description:Quick-DNA™ Viral 96 Kit