Order Entry
Canada
ContactUsLinkComponent
Quick-DNA™ Tissue/Insect Kits, Zymo Research
Quick-DNA™ Tissue/Insect Kits, Zymo Research
Supplier:  Zymo Research
Certificates
undefined
Quick-DNA™ Tissue/Insect Kits, Zymo Research
Supplier:  Zymo Research
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Cultured cells
    Invertebrates
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Description:
    Quick-DNA™ Tissue/Insect Miniprep Kit

Specifications

Frequently Bought Together