Order Entry
Canada
ContactUsLinkComponent
Epitope tag
Epitope tag
Catalog # 103003-362
Supplier:  Anaspec Inc
CAS Number:  
Epitope tag
Catalog # 103003-362
Supplier:  Anaspec Inc
Supplier Number:  AS-60738
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    5 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Hydrophilic epitope tag|DYKDDDK tag
  • Protein/peptide name:
    Epitope tag
  • Purity:
    95%
  • Molecular weight:
    1013 Da
  • Sequence:
    DYKDDDDK
  • Cat. no.:
    103003-362
  • Supplier No.:
    AS-60738

Specifications

About this item

This epitope tag is a short hydrophilic, highly charged peptide. It is the most widely used epitope tag employed in structural and functional studies of proteins.
Sequence:DYKDDDDK
MW:1013 Da
% peak area by HPLC:95
Storage condition:-20° C