This is a hexapeptide with 6 histidines primarily used in tagging proteins. His tags have been used in affinity purification of recombinant proteins and have also been used in studying protein transduction in cells with the use of cell-penetrating proteins/peptides.
Sequence:HHHHHH
MW:841.9 Da
% peak area by HPLC:95
Storage condition:-20° C
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Storage conditions:–20 °C
- Protein synonyms:Affinity tag|Epitope tag
- Protein/peptide name:Hexa His
- Purity:95%
- Molecular weight:841.9 Da
- Sequence:HHHHHH