Order Entry
Canada
ContactUsLinkComponent
SV-40 Large T-Antigen Nuclear Localization Signal
SV-40 Large T-Antigen Nuclear Localization Signal
Catalog # 103007-760
Supplier:  Anaspec Inc
CAS Number:  
SV-40 Large T-Antigen Nuclear Localization Signal
Catalog # 103007-760
Supplier:  Anaspec Inc
Supplier Number:  AS-63831
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Large T antigen Peptide|Nuclear translocation Peptide|DNA tag Peptide|NLS Peptide|Nuclear localization ppetide
  • Protein/peptide name:
    SV-40 Large T-antigen Nuclear Localization Signal
  • Purity:
    95%
  • Molecular weight:
    1401.7 Da
  • Sequence:
    CGGGPKKKRKVED
  • Cat. no.:
    103007-760
  • Supplier No.:
    AS-63831

Specifications

About this item

This peptide is derived from Large T antigen residue 47 to 55 (PKKKRKVED). It is a commonly used nuclear localization signal (NLS) peptide. It enables protein import into cell nucleus.
Sequence:CGGGPKKKRKVED
MW:1401.7 Da
% peak area by HPLC:95
Storage condition:-20° C