Order Entry
Canada
ContactUsLinkComponent
SV40 T-Ag-derived Nuclear Localization Signal (NLS) Peptide
SV40 T-Ag-derived Nuclear Localization Signal (NLS) Peptide
Catalog # 103007-728
Supplier:  Anaspec Inc
CAS Number:  
SV40 T-Ag-derived Nuclear Localization Signal (NLS) Peptide
Catalog # 103007-728
Supplier:  Anaspec Inc
Supplier Number:  AS-63788
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Large T antigen Peptide|Nuclear translocation Peptide|DNA tag Peptide|NLS Peptide|Nuclear localization ppetide
  • Protein/peptide name:
    SV40 T-Ag-derived Nuclear Localization Signal (NLS) Peptide
  • Purity:
    95%
  • Molecular weight:
    1490.8 Da
  • Sequence:
    PKKKRKVEDPYC
  • Cat. no.:
    103007-728
  • Supplier No.:
    AS-63788

Specifications

About this item

This peptide, a nuclear localization signal (NLS) peptide, is derived from the Large T antigen residues 47 to 56 (PKKKRKVEDP). It can be used to tag DNA. DNA tagged to this peptide efficiently translocates into the cell nucleus.
Sequence:PKKKRKVEDPYC
MW:1490.8 Da
% peak area by HPLC:95
Storage condition:-20° C