Order Entry
Canada
ContactUsLinkComponent
 
Drosophila Antennapedia Peptide, FITC (Fluorescein Isothiocyanate)
 
Drosophila Antennapedia Peptide, FITC (Fluorescein Isothiocyanate)

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Species:
    Drosophila
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Antennapedia homeodomain
  • Protein/peptide name:
    Antennapedia Peptide
  • Purity:
    95%
  • Molecular weight:
    2748.3 Da
  • Sequence:
    FITC-LC-RQIKIWFQNRRMKWKK-NH2

 

Frequently Bought Together