Order Entry
Canada
ContactUsLinkComponent
Interphotoreceptor Retinoid Binding Protein Fragment
Interphotoreceptor Retinoid Binding Protein Fragment
Catalog # 103002-994
Supplier:  Anaspec Inc
CAS Number:  
Interphotoreceptor Retinoid Binding Protein Fragment
Catalog # 103002-994
Supplier:  Anaspec Inc
Supplier Number:  AS-60214-5
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    5 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    K18 Peptide|Keratin Peptide
  • Protein/peptide name:
    Interphotoreceptor Retinoid Binding Protein Fragment
  • Purity:
    95%
  • Molecular weight:
    1418.5 Da
  • Sequence:
    RPVSSAApSVYAGAC
  • Cat. no.:
    103002-994
  • Supplier No.:
    AS-60214-5

Specifications

About this item

This peptide corresponds to human keratin K18 amino acids 26-38, with an additional C-terminal cysteine. The sequence is identical to mouse K18 except that human K18 Val28 is replaced by alanine in the mouse sequence. Members of the 14-3-3 protein family bind the human intermediate filament protein keratin 18 (K18) in-vivo, in a cell-cycle- and phosphorylation-dependent manner. K18 Ser33 phosphorylation is essential for the association of K18 with 14-3-3 proteins and plays a role in keratin organization and distribution.
Sequence:RPVSSAApSVYAGAC
MW:1418.5 Da
% peak area by HPLC:95
Storage condition:-20° C