Order Entry
Canada
ContactUsLinkComponent
Interphotoreceptor Retinoid Binding Protein Fragment Peptide
Interphotoreceptor Retinoid Binding Protein Fragment Peptide
Catalog # 103008-232
Supplier:  Anaspec Inc
CAS Number:  
Interphotoreceptor Retinoid Binding Protein Fragment Peptide
Catalog # 103008-232
Supplier:  Anaspec Inc
Supplier Number:  AS-64549
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Interphotoreceptor retinoid binding protein derived Peptide
  • Protein/peptide name:
    Interphotoreceptor Retinoid Binding Protein Fragment Peptide
  • Purity:
    95%
  • Molecular weight:
    1473.6 Da
  • Sequence:
    ADGSSWEGVGVVPDV
  • Cat. no.:
    103008-232
  • Supplier No.:
    AS-64549

Specifications

About this item

R16 is an IRBP (Interphotoreceptor retinoid binding protein) derived peptide. Photoreceptor cell protein is capable of inducing an experimental autoimmune uveitis (EAU) in susceptible animal strains.
Sequence:ADGSSWEGVGVVPDV
MW:1473.6 Da
% peak area by HPLC:95
Storage condition:-20° C