Order Entry
Canada
ContactUsLinkComponent
Dipeptidylaminopeptidase IV Substrate, AMC (7-amino-4-methylcoumarin)
Dipeptidylaminopeptidase IV Substrate, AMC (7-amino-4-methylcoumarin)
Catalog # 102996-430
Supplier:  Anaspec Inc
CAS Number:  
Dipeptidylaminopeptidase IV Substrate, AMC (7-amino-4-methylcoumarin)
Catalog # 102996-430
Supplier:  Anaspec Inc
Supplier Number:  AS-24098
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    AMC (7-amino-4-methylcoumarin)
  • Size:
    10 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    DPP-IV substrate
  • Protein/peptide name:
    Dipeptidylaminopeptidase IV Substrate
  • Purity:
    95%
  • Molecular weight:
    329.2 Da
  • Sequence:
    GP-AMC
  • Cat. no.:
    102996-430
  • Supplier No.:
    AS-24098

Specifications

About this item

This is a fluorescent dipeptidylaminopeptidase IV substrate, Abs/Em=353/442 nm.
Sequence:GP-AMC
MW:329.2 Da
% peak area by HPLC:95
Storage condition:-20° C