Kallikrein 3 Substrate
Catalog # 103003-896
Supplier: Anaspec Inc
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Size:5 mg
- Storage conditions:–20 °C
- Protein synonyms:Kallikrin substrate|PSA substrate
- Protein/peptide name:Kallikrein 3 Substrate
- Purity:95%
- Molecular weight:654.6 Da
- Sequence:Suc-RPY-pNA
- Cat. no.:103003-896
- Supplier No.:AS-20586
Specifications
About this item
Chromogenic substrate for Kallikrein 3, commonly known as prostate specific antigen (PSA)
Sequence:Suc-RPY-pNA
MW:654.6 Da
% peak area by HPLC:95
Storage condition:-20° C