Order Entry
Canada
ContactUsLinkComponent
Tyrosine Kinase Peptide 1, TMR (Tetramethylrhodamine)
Tyrosine Kinase Peptide 1, TMR (Tetramethylrhodamine)
Catalog # 102997-410
Supplier:  Anaspec Inc
CAS Number:  
Tyrosine Kinase Peptide 1, TMR (Tetramethylrhodamine)
Catalog # 102997-410
Supplier:  Anaspec Inc
Supplier Number:  AS-29935-1
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    TMR (Tetramethylrhodamine)
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Tyrosine protein kinase substrate
  • Protein/peptide name:
    Tyrosine Kinase Peptide 1
  • Purity:
    95%
  • Molecular weight:
    2082.5 Da
  • Sequence:
    5-TMR-KVEKIGEGTYGVVYK
  • Cat. no.:
    102997-410
  • Supplier No.:
    AS-29935-1

Specifications

About this item

Peptide sequence KVEKIGEGTYGVVYK is derived from the amino acid residues CDC26-20. It is considered to be a generic substrate for various TPKs.
Sequence:5-TMR-KVEKIGEGTYGVVYK
MW:2082.5 Da
% peak area by HPLC:95
Storage condition:-20° C