Order Entry
Canada
ContactUsLinkComponent
Src Optimal Peptide Substrate
Src Optimal Peptide Substrate
Catalog # 103003-006
Supplier:  Anaspec Inc
CAS Number:  
Src Optimal Peptide Substrate
Catalog # 103003-006
Supplier:  Anaspec Inc
Supplier Number:  AS-60222-1
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Src substrate
  • Protein/peptide name:
    Src Optimal Peptide Substrate
  • Purity:
    95%
  • Molecular weight:
    1799 Da
  • Sequence:
    AEEEIYGEFEAKKKK
  • Cat. no.:
    103003-006
  • Supplier No.:
    AS-60222-1

Specifications

About this item

The synthetic peptide has been identified as a Src optimal peptide substrate. It may be used to measure the wild type Src activity in comparison with other family members.
Sequence:AEEEIYGEFEAKKKK
MW:1799 Da
% peak area by HPLC:95
Storage condition:-20° C