Order Entry
Canada
ContactUsLinkComponent
pp60(v-SRC) Autophosphorylation Site
pp60(v-SRC) Autophosphorylation Site
Catalog # 102996-272
Supplier:  Anaspec Inc
CAS Number:  
pp60(v-SRC) Autophosphorylation Site
Catalog # 102996-272
Supplier:  Anaspec Inc
Supplier Number:  AS-22555
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Rous sarcoma virus pp60 tyrosine phosphorylation Peptide|pp60 SRC Peptide.
  • Protein/peptide name:
    pp60(v-SRC) Autophosphorylation Site
  • Purity:
    95%
  • Molecular weight:
    1672.7 Da
  • Sequence:
    RRLIEDNE-pY-TARG
  • Cat. no.:
    102996-272
  • Supplier No.:
    AS-22555

Specifications

About this item

These sequences correspond to the tyrosine phosphorylation site of the Rous sarcoma virus-encoded transforming protein pp60 SRC.
Sequence:RRLIEDNE-pY-TARG
MW:1672.7 Da
% peak area by HPLC:95
Storage condition:-20° C