Order Entry
Canada
ContactUsLinkComponent
Syk Kinase Peptide Substrate
Syk Kinase Peptide Substrate
Catalog # 103007-866
Supplier:  Anaspec
CAS Number:  
Syk Kinase Peptide Substrate
Catalog # 103007-866
Supplier:  Anaspec
Supplier Number:  AS-64141
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Syk kinase substrate
  • Protein/peptide name:
    Syk Kinase Peptide Substrate
  • Purity:
    95%
  • Molecular weight:
    1463.6 Da
  • Sequence:
    KEDPDYEWPSAK-NH2
  • Cat. no.:
    103007-866
  • Supplier No.:
    AS-64141

Specifications

About this item

This synthetic peptide is a substrate for Syk kinase.
Sequence:KEDPDYEWPSAK-NH2
MW:1463.6 Da
% peak area by HPLC:95
Storage condition:-20° C