Order Entry
Canada
ContactUsLinkComponent
S3 Fragment, ADF/cofilin
S3 Fragment, ADF/cofilin
Catalog # 103007-442
Supplier:  Anaspec Inc
CAS Number:  
S3 Fragment, ADF/cofilin
Catalog # 103007-442
Supplier:  Anaspec Inc
Supplier Number:  AS-62637
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    LIM kinase inhibitor|LIMK1 inhibitor Peptide
  • Protein/peptide name:
    S3 Fragment, ADF/cofilin
  • Purity:
    95%
  • Molecular weight:
    1698 Da
  • Sequence:
    MASGVMVSDDVVKVFN
  • Cat. no.:
    103007-442
  • Supplier No.:
    AS-62637

Specifications

About this item

This peptide contains the unique amino-terminal phosphorylation site of Xenopus ADF/cofilin, the LIM kinase (LIMK) phosphorylation site. LIMK1 is a key regulator of the actin cytoskeleton through its phosphorylation of ADF/cofilin at serine-3 for inactivation. This peptide is a fragment of the S3 peptide containing the serine-3 sequence of ADF/cofilin that has been widely used as an effective competitive inhibitor of LIMK1.
Sequence:MASGVMVSDDVVKVFN
MW:1698 Da
% peak area by HPLC:95
Storage condition:-20° C