Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Size:1 mg
- Storage conditions:–20 °C
- Protein synonyms:Abl Kinase substrate|Abl protein tyrosine kinase substrate
- Protein/peptide name:Abltide
- Purity:95%
- Molecular weight:1264.5 Da
- Sequence:KKGEAIYAAPFA-NH2
- Cat. no.:103006-960
- Supplier No.:AS-61749
Abltide is a peptide substrate for Abl Kinase (Abl protein tyrosine kinase), a partner in the gag-Abl fusion protein of the Abelson murine leukemia virus. Used in Western blot and kinase assay.
Sequence:KKGEAIYAAPFA-NH2
MW:1264.5 Da
% peak area by HPLC:95
Storage condition:-20° C