Order Entry
Canada
ContactUsLinkComponent
IRS1-Derived Peptide
IRS1-Derived Peptide
Catalog # 103006-974
Supplier:  Anaspec Inc
CAS Number:  
IRS1-Derived Peptide
Catalog # 103006-974
Supplier:  Anaspec Inc
Supplier Number:  AS-61764
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    PI3 kinase binding domain Peptide|Insulin receptor substrate-1 Peptide
  • Protein/peptide name:
    IRS1-derived Peptide
  • Purity:
    95%
  • Molecular weight:
    1513.8 Da
  • Sequence:
    KKSRGDYMTMQIG-NH2
  • Cat. no.:
    103006-974
  • Supplier No.:
    AS-61764

Specifications

About this item

This is a peptide fragment (979-989) of the insulin receptor substrate-1 containing the sequence motif YMXM known to bind to the two domains of SH2 on the 85kDa subunit of phosphoinositide 3-kinase.
Sequence:KKSRGDYMTMQIG-NH2
MW:1513.8 Da
% peak area by HPLC:95
Storage condition:-20° C