Order Entry
Canada
ContactUsLinkComponent
GSK3 Substrate, a, b Subunit
GSK3 Substrate, a, b Subunit
Catalog # 103003-268
Supplier:  Anaspec
CAS Number:  
GSK3 Substrate, a, b Subunit
Catalog # 103003-268
Supplier:  Anaspec
Supplier Number:  AS-60617
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Glycogen synthase kinase 3 substrate
  • Protein/peptide name:
    GSK3 Substrate, a, b subunit
  • Purity:
    95%
  • Molecular weight:
    2631.8 Da
  • Sequence:
    RAAVPPSPSLSRHSSPHQSEDEEE
  • Cat. no.:
    103003-268
  • Supplier No.:
    AS-60617

Specifications

About this item

This is a GSK-3 substrate, it can be used as a substrate for both alpha and beta isoforms of GSK3.
Sequence:RAAVPPSPSLSRHSSPHQSEDEEE
MW:2631.8 Da
% peak area by HPLC:95
Storage condition:-20° C