Cyclin-dependent protein kinase 1 Substrate
Catalog # 103003-204
Supplier: Anaspec Inc
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Size:5 mg
- Storage conditions:–20 °C
- Protein synonyms:CDK1 substrate|cyclin dependent kinase 1 substrate
- Protein/peptide name:Cyclin-dependent protein kinase 1 Substrate
- Purity:95%
- Molecular weight:1190.5 Da
- Sequence:HATPPKKKRK
- Cat. no.:103003-204
- Supplier No.:AS-60522-5
Specifications
About this item
The native peptide, HATPPKKKRK (cat# 60522-1), is a substrate for cyclin-dependent protein kinase 1 (CDC2; CDK1). The fluorescent and biotinylated peptides are used to develop assays for CDK1.
Sequence:HATPPKKKRK
MW:1190.5 Da
% peak area by HPLC:95
Storage condition:-20° C