Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Size:1 mg
- Storage conditions:–20 °C
- Protein synonyms:Casein Kinase 2 substrate|CK2 substrate
- Protein/peptide name:Casein Kinase 2 (CK2) Substrate alpha
- Purity:95%
- Molecular weight:1264.2 Da
- Sequence:RRRDDDSDDD
- Cat. no.:103003-266
- Supplier No.:AS-60615
Specifications
About this item
Type II casein kinase (CK-2) is unique among the protein kinases since it can use ATP as well as GTP as a phosphoryl donor. It is extremely sensitive to heparin inhibition and can be activated by polyamines and basic polypeptides. This peptide contains the consensus phosphorylation sequence for CK2 and can be used as the substrate for CK2 a-subunit.
Sequence:RRRDDDSDDD
MW:1264.2 Da
% peak area by HPLC:95
Storage condition:-20° C