Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Size:1 mg
- Storage conditions:–20 °C
- Protein synonyms:AIP|Myristoylated Autocatamide inhibitory Peptide|Myristoylated AIP|CaMKII inhibitor substrate|Calmodulin-Dependent Protein Kinase ll inhibitor substrate
- Protein/peptide name:Autocamtide-2-Related Inhibitory Peptide
- Purity:95%
- Molecular weight:1708.2 Da
- Sequence:Myr-KKALRRQEAVDAL
- Cat. no.:103008-588
- Supplier No.:AS-64929
This is a myristoylated form of Autocamtide-2-Related Inhibitory Peptide (AIP), a highly potent and specific substrate competitive inhibitor of Calmodulin-Dependent Protein Kinase ll (CaMKII). AIP is derived from Autocamtide-2, a substrate for CaMKII, with the Thr-9 phosphorylation site substituted with Ala.
Sequence:Myr-KKALRRQEAVDAL
MW:1708.2 Da
% peak area by HPLC:95
Storage condition:-20° C