About this item
The native peptide KKALRRQETVDAL (60249-1) is a selective substrate for Ca2+/calmodulin-dependent protein kinase (CaMK). The fluorescent and biotinylated peptides are used to design assays for CaMKs.
Sequence:KKALRRQETVDAL
MW:1527.8 Da
% peak area by HPLC:95
Storage condition:-20° C
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Storage conditions:–20 °C
- Protein synonyms:Ca2+/calmodulin-dependent protein kinase substrate|CaM kinase substrate|CaMK substrate
- Protein/peptide name:Autocatamide-2 Peptide
- Purity:95%
- Molecular weight:1527.8 Da
- Sequence:KKALRRQETVDAL