About this item
The native peptide ARKRERTYSFGHHA (29944-1) is a synthetic substrate for AKT/PKB/Rac-protein kinases. Phophorylation is at the Ser site (Km = 3.9 µM). It also competitively inhibits histone H2B phosphorylation (Ki = 12 µM) by AKT.
Sequence:Biotin-ARKRERTYSFGHHA
MW:1942.2 Da
% peak area by HPLC:95
Storage condition:-20° C
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Biotin
- Storage conditions:–20 °C
- Protein synonyms:AKT/PKB/Rac Kinase Substrate|Kinase substrate|histone H2B phosphorylation inhibitor
- Protein/peptide name:AKT/PKB/Rac-Protein Kinase Substrate
- Purity:95%
- Molecular weight:1942.2 Da
- Sequence:Biotin-ARKRERTYSFGHHA