Order Entry
Canada
ContactUsLinkComponent
Histone Deacetylase Substrate, AMC (7-Amino-4-Methylcoumarin)
Histone Deacetylase Substrate, AMC (7-Amino-4-Methylcoumarin)
Catalog # 103007-068
Supplier:  Anaspec Inc
CAS Number:  
Histone Deacetylase Substrate, AMC (7-Amino-4-Methylcoumarin)
Catalog # 103007-068
Supplier:  Anaspec Inc
Supplier Number:  AS-61855
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    AMC (7-amino-4-methylcoumarin)
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Fluorogenic HDAC substrate
  • Protein/peptide name:
    Histone deacetylase Substrate
  • Purity:
    95%
  • Molecular weight:
    600.7 Da
  • Sequence:
    Ac-RGK(Ac)-AMC
  • Cat. no.:
    103007-068
  • Supplier No.:
    AS-61855

Specifications

About this item

This novel fluorogenic substrate of HDACs was synthesized with an epsilon-acetylated lysyl moiety and an adjacent MCA moiety at the C-terminus of the peptide chain. The assay utilizing this substrate provides a good tool to characterize the HDAC activity. HDACs are important enzymes for the transcriptional regulation of gene expression.
Sequence:Ac-RGK(Ac)-AMC
MW:600.7 Da
% peak area by HPLC:95
Storage condition:-20° C