Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Biotin
- Size:1 mg
- Storage conditions:–20 °C
- Protein synonyms:Histone H4 Peptide|H4K12/16
- Protein/peptide name:[Lys(Ac)12/16]-Histone H4(1-25)-GSGSK
- Purity:95%
- Molecular weight:3316.8 Da
- Sequence:SGRGKGGKGLG-K(Ac)-GGA-K(Ac)-RHRKVLRDN-GSGSK(Biotin)
- Cat. no.:103009-070
- Supplier No.:AS-65222-1
Specifications
About this item
This peptide is histone H4 (1-25) with acetylation at Lys12 and Lys16. It is biotinylated through a C-terminal GSGSK linker. It has been shown that human histone H4 is preferentially acetylated first at Lys16, then at Lys12. Diacetylated histone H4 has been shown to bind repressor protein TUP1. Acetylation of histone H4 also regulates heterochromatin formation by promoting the binding of bromodomains to p300 and transcription factor TAFII250 and inhibiting interactions with SIR3. Provided at >95% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Sequence:SGRGKGGKGLG-K(Ac)-GGA-K(Ac)-RHRKVLRDN-GSGSK(Biotin)
MW:3316.8 Da
% peak area by HPLC:95
Storage condition:-20° C