Order Entry
Canada
ContactUsLinkComponent
[Arg(Me1)3]-Histone H4 (1-23)-GGK,Biotin
[Arg(Me1)3]-Histone H4 (1-23)-GGK,Biotin
Catalog # 103009-358
Supplier:  Anaspec Inc
CAS Number:  
[Arg(Me1)3]-Histone H4 (1-23)-GGK,Biotin
Catalog # 103009-358
Supplier:  Anaspec Inc
Supplier Number:  AS-65277-1
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Biotin
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Histone H4 Peptide
  • Protein/peptide name:
    [Arg(Me1)3]-Histone H4 (1-23)-GGK
  • Purity:
    95%
  • Molecular weight:
    2843.4 Da
  • Sequence:
    SG-R(Me1)-GKGGKGLGKGGAKRHRKVLR-GGK(Biotin)
  • Cat. no.:
    103009-358
  • Supplier No.:
    AS-65277-1

Specifications

About this item

This peptide is histone H4 (1-23) monomethylated at Arg3 followed by a biotinylated Lys conjugated to a C-terminal GG linker. Methylation of Arg3 is catalyzed by PRMT1 and functions to promote p300 acetylation of histone H4. Provided at >95% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Sequence:SG-R(Me1)-GKGGKGLGKGGAKRHRKVLR-GGK(Biotin)
MW:2843.4 Da
% peak area by HPLC:95
Storage condition:-20° C