Histone H4 (1-20) Substrate
Catalog # 103007-370
Supplier: Anaspec Inc
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Size:1 mg
- Storage conditions:–20 °C
- Protein synonyms:PRMT7|Histone H4 Peptide
- Protein/peptide name:Histone H4 (1-20) Substrate
- Purity:95%
- Molecular weight:1991.3 Da
- Sequence:SGRGKGGKGLGKGGAKRHRK-NH2
- Cat. no.:103007-370
- Supplier No.:AS-62498
Specifications
About this item
This GRG containing peptide is amino acids 1 to 20 fragment of histone 4. It is a substrate for human protein-arginine methyltransferase 7 (PRMT7), a type II methyltransferase capable of producing symmetrical dimethyl arginine (sDMA) modifications in proteins.
Sequence:SGRGKGGKGLGKGGAKRHRK-NH2
MW:1991.3 Da
% peak area by HPLC:95
Storage condition:-20° C