Order Entry
Canada
ContactUsLinkComponent
[Lys(Me1)36]-Histone H3 (31-41)
[Lys(Me1)36]-Histone H3 (31-41)
Catalog # 103008-036
Supplier:  Anaspec Inc
CAS Number:  
[Lys(Me1)36]-Histone H3 (31-41)
Catalog # 103008-036
Supplier:  Anaspec Inc
Supplier Number:  AS-64379-1
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Histone H3 Peptide|H3K36
  • Protein/peptide name:
    [Lys(Me1)36]-Histone H3 (31-41)
  • Purity:
    95%
  • Molecular weight:
    1243.4 Da
  • Sequence:
    STGGV-K(Me1)-KPHRY
  • Cat. no.:
    103008-036
  • Supplier No.:
    AS-64379-1

Specifications

About this item

This peptide is Histone H3 amino acid residues 31 to 41 mono-methylated at Lys-36.
Sequence:STGGV-K(Me1)-KPHRY
MW:1243.4 Da
% peak area by HPLC:95
Storage condition:-20° C