Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Size:1 mg
- Storage conditions:–20 °C
- Protein synonyms:Histone H3 Peptide|H3K27
- Protein/peptide name:[Lys(Me1)27]-Histone H3 (23-34)
- Purity:95%
- Molecular weight:1128.3 Da
- Sequence:KAAR-K(Me1)-SAPATGG
- Cat. no.:103008-030
- Supplier No.:AS-64376-1
This peptide is Histone H3 amino acid residues 23 to 34 mono-methylated at Lys-27.
Sequence:KAAR-K(Me1)-SAPATGG
MW:1128.3 Da
% peak area by HPLC:95
Storage condition:-20° C