Histone H2B (21-41), Biotin
Catalog # 103008-048
Supplier: Anaspec Inc
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Biotin
- Size:1 mg
- Storage conditions:–20 °C
- Protein synonyms:Histone 2B Peptide
- Protein/peptide name:Histone H2B (21-41)
- Purity:95%
- Molecular weight:3024.6 Da
- Sequence:AQKKDGKKRKRSRKESYSIYV-GGK(Biotin)
- Cat. no.:103008-048
- Supplier No.:AS-64385-1
Specifications
About this item
This peptide is Histone 2B amino acid residues 21 to 41 with a C-terminal GG linker followed by a biotinylated lysine.
Sequence:AQKKDGKKRKRSRKESYSIYV-GGK(Biotin)
MW:3024.6 Da
% peak area by HPLC:95
Storage condition:-20° C