Order Entry
Canada
ContactUsLinkComponent
Shepherdin (79–87)
Shepherdin (79–87)
Catalog # 103007-504
Supplier:  Anaspec Inc
CAS Number:  
Shepherdin (79–87)
Catalog # 103007-504
Supplier:  Anaspec Inc
Supplier Number:  AS-62748
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Shepherdin Peptide
  • Protein/peptide name:
    Shepherdin (79–87)
  • Purity:
    95%
  • Molecular weight:
    949.1 Da
  • Sequence:
    KHSSGCAFL
  • Cat. no.:
    103007-504
  • Supplier No.:
    AS-62748

Specifications

About this item

This is amino acids 79 to 87 fragment of shepherdin, a novel peptidomimetic antagonist of the complex between Hsp90 and survivin, another key regulator of tumor cell viability. For its potent and broad antitumor activity, selectivity of action in tumor cells versus normal tissues, and inhibition of tumor growth in vivo without toxicity, shepherdin may offer a promising approach for rational cancer therapy. This sequence is also known as K79–K90.
Sequence:KHSSGCAFL
MW:949.1 Da
% peak area by HPLC:95
Storage condition:-20° C