Order Entry
Canada
ContactUsLinkComponent
HIF-1 {alpha} (556-574)
HIF-1 {alpha} (556-574)
Catalog # 103006-808
Supplier:  Anaspec Inc
CAS Number:  
HIF-1 {alpha} (556-574)
Catalog # 103006-808
Supplier:  Anaspec Inc
Supplier Number:  AS-61528
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    HIF-1 a Peptide|HIF-1 19 mer|hypoxia-inducible factor-1 Peptide
  • Protein/peptide name:
    HIF-1 {alpha} (556-574)
  • Purity:
    95%
  • Molecular weight:
    2254.6 Da
  • Sequence:
    DLDLEMLAPYIPMDDDFQL
  • Cat. no.:
    103006-808
  • Supplier No.:
    AS-61528

Specifications

About this item

This is a hypoxia-inducible factor-1 (HIF-1 a) 19-mer fragment. HIF-1 functions as master regulator of response to oxygen homeostasis. Hypoxia-induced gene expression is initiated when HIF-1 subunit is stabilized in response to a lack of oxygen. This part of HIF-1 binds to the von Hippel-Lindau factor (VHL) an E3 ubiquitin ligase, and the proline 564 is absolutely critical to the binding process
Sequence:DLDLEMLAPYIPMDDDFQL
MW:2254.6 Da
% peak area by HPLC:95
Storage condition:-20° C