Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Size:1 mg
- Storage conditions:–20 °C
- Protein synonyms:p53N|Mdm-2 binding P53 domain
- Protein/peptide name:p53 (17-26)
- Purity:95%
- Molecular weight:1251.5 Da
- Sequence:ETFSDLWKLL
- Cat. no.:103006-594
- Supplier No.:AS-61375
This peptide is amino acids 17 to 26 fragment of p53, the Mdm-2 binding domain of p53 known also as p53N. This sequence contains all of the residues that contact the binding domain of Mdm-2. The tumor suppressor protein p53 is important in maintaining genome stability and in preventing cancer development.
Sequence:ETFSDLWKLL
MW:1251.5 Da
% peak area by HPLC:95
Storage condition:-20° C