Order Entry
Canada
ContactUsLinkComponent
Proapoptotic Peptide
Proapoptotic Peptide
  103007-236
 :  
Proapoptotic Peptide
  103007-236
 :  AS-62199
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    klaklak Peptide
  • Protein/peptide name:
    Proapoptotic Peptide
  • Purity:
    95%
  • Molecular weight:
    1523 Da
  • Sequence:
    klaklakklaklak-NH2
  • Cat. no.:
    103007-236
  • Supplier No.:
    AS-62199

 

 

This pro-apoptotic peptide, composed of D-amino acids, is a alpha-helical amphipathic peptide toxic to eukaryotic cells if internalized by a suitable targeting mechanism. It disrupts mitochondrial membranes upon receptor-mediated cell internalization and causes programmed cell death.
Sequence:klaklakklaklak-NH2
MW:1523 Da
% peak area by HPLC:95
Storage condition:-20° C