Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Species:Cytomegalovirus (CMV)
- Size:1 mg
- Storage conditions:–20 °C
- Protein synonyms:CMV matrix phosphoprotein Peptide
- Protein/peptide name:CMV pp65 Peptide
- Purity:95%
- Molecular weight:1523.8 Da
- Sequence:SVLGPISGHVLKAVF
- Cat. no.:103007-816
- Supplier No.:AS-63937
This peptide is derived from amino acid residues 13 to 27 of the 65k lower matrix phosphoprotein of the human cytomegalovirus.It contains a nine-amino-acid sequence (LGPISGHVL) that matches the consensus binding motif for a major histocompatibility complex H2-Dd T-cell epitope.
Sequence:SVLGPISGHVLKAVF
MW:1523.8 Da
% peak area by HPLC:95
Storage condition:-20° C
Sequence:SVLGPISGHVLKAVF
MW:1523.8 Da
% peak area by HPLC:95
Storage condition:-20° C