Order Entry
Canada
ContactUsLinkComponent
Staphylococcus aureus Delta-Toxin (1-26)
Staphylococcus aureus Delta-Toxin (1-26)
Catalog # 103007-368
Supplier:  Anaspec Inc
CAS Number:  
Staphylococcus aureus Delta-Toxin (1-26)
Catalog # 103007-368
Supplier:  Anaspec Inc
Supplier Number:  AS-62496
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Staphylococcus aureus
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Delta-lysin|hemolytic Peptide
  • Protein/peptide name:
    Delta-Toxin (1-26)
  • Purity:
    95%
  • Molecular weight:
    2978.6 Da
  • Sequence:
    MAQDIISTIGDLVKWIIDTVNKFTKK
  • Cat. no.:
    103007-368
  • Supplier No.:
    AS-62496

Specifications

About this item

Delta -Lysin is a 26-residue hemolytic peptide secreted by Staphylococcus aureus, it is a delta-toxin. It is produced by most of the strains of S. aureus. This toxin is lytic to many kinds of cells by solubilization of their membranes.
Sequence:MAQDIISTIGDLVKWIIDTVNKFTKK
MW:2978.6 Da
% peak area by HPLC:95
Storage condition:-20° C