Order Entry
Canada
ContactUsLinkComponent
Human HNP-1 Peptide
Human HNP-1 Peptide
  103003-370
 :  
Human HNP-1 Peptide
  103003-370
 :  AS-60743
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Size:
    0.1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    human neutrophil Peptide 2|human a -defensin
  • Protein/peptide name:
    HNP-1 Peptide
  • Purity:
    95%
  • Molecular weight:
    3442.1 Da
  • Sequence:
    ACYCRIPACIAGERRYGTCIYQGRLWAFCC (Disulfide bridge: 2-30, 4-19, 9-29)
  • Cat. no.:
    103003-370
  • Supplier No.:
    AS-60743

 

 

Mammalian defensins are abundant in the cytoplasmic azurophilic granules of neutrophils, Paneth cells of the small intestine and some macrophages. Human a-defensin-1 (HNP-1) is a peptide possessing both broad antimicrobial (both Gram-positive and Gram-negative bacteria) and cytotoxic activities. HNP-1 reduces adenoviral infection by more than 95%.
Sequence:ACYCRIPACIAGERRYGTCIYQGRLWAFCC (Disulfide bridge: 2-30, 4-19, 9-29)
MW:3442.1 Da
% peak area by HPLC:95
Storage condition:-20° C