Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Species:Sheep
- Size:1 mg
- Storage conditions:–20 °C
- Protein synonyms:sheep myeloid Peptide
- Protein/peptide name:SMAP 29 Peptide
- Purity:95%
- Molecular weight:3256 Da
- Sequence:RGLRRLGRKIAHGVKKYGPTVLRIIRIAG
- Cat. no.:103006-564
- Supplier No.:AS-61308
Specifications
About this item
This 29 amino acid sheep myeloid antimicrobial peptide 29 (SMAP 29) displays extremely high antimicrobial activity against Pseudomonas strains, other gram-negative bacteria, and multidrug-resistant pathogens. SMAP 29 is a broad-spectrum antibiotic cathelicidin, it is active in both low- and high-ionic-strength conditions, and induces significant morphological alterations in bacterial surfaces.
Sequence:RGLRRLGRKIAHGVKKYGPTVLRIIRIAG
MW:3256 Da
% peak area by HPLC:95
Storage condition:-20° C