Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Species:Rat
- Size:0.5 mg
- Storage conditions:–20 °C
- Protein synonyms:CRAMP|rat cathelicidin Peptide
- Protein/peptide name:rCRAMP
- Purity:95%
- Molecular weight:3946.7 Da
- Sequence:GLVRKGGEKFGEKLRKIGQKIKEFFQKLALEIEQ
- Cat. no.:103006-560
- Supplier No.:AS-61306
This peptide is a rat homologue of the human cathelicidin LL-37. Rat cathelicidin-related antimicrobial peptide (rCRAMP) was identified in granulocytes, thymus, testis, lung, mouth mucosa, tongue, oesophagus, colon, caecum and small intestine. The rCRAMP peptide is present in specific CNS regions and may play a role in the innate immunity of the CNS. rCRAMP exhibits pro-healing activity in stomach.
Sequence:GLVRKGGEKFGEKLRKIGQKIKEFFQKLALEIEQ
MW:3946.7 Da
% peak area by HPLC:95
Storage condition:-20° C