Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Species:Mouse
- Size:1 mg
- Storage conditions:–20 °C
- Protein synonyms:Complement component Peptide|C2 DNP|C2 Peptide|Cls Substrate Peptide
- Protein/peptide name:Cls Substrate
- Purity:95%
- Molecular weight:1326.5 Da
- Sequence:2Abz-SLGRKIQIK(Dnp)-NH2
- Cat. no.:103006-570
- Supplier No.:AS-61315
This peptide is a second complement component (C2), the physiological substrate for the proenzyme Cls, first complement component. The complement system is a central component of host defense but can also contribute to the inflammation seen in pathological conditions. The C1s protease of the C1 complex initiates the host defense pathway. This peptide employs 2Abz/Dnp FRET pair for quantitation of complement enzyme activity.
Sequence:2Abz-SLGRKIQIK(Dnp)-NH2
MW:1326.5 Da
% peak area by HPLC:95
Storage condition:-20° C