Order Entry
Canada
ContactUsLinkComponent
Mouse Neuropeptide S
Mouse Neuropeptide S
Catalog # 103006-452
Supplier:  Anaspec Inc
CAS Number:  
Mouse Neuropeptide S
Catalog # 103006-452
Supplier:  Anaspec Inc
Supplier Number:  AS-61246
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Mouse
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    NPS Peptide
  • Protein/peptide name:
    Neuropeptide S
  • Purity:
    95%
  • Molecular weight:
    2182.5 Da
  • Sequence:
    SFRNGVGSGAKKTSFRRAKQ
  • Cat. no.:
    103006-452
  • Supplier No.:
    AS-61246

Specifications

About this item

A neuropeptide that plays a role in arousal and fear responses.
Sequence:SFRNGVGSGAKKTSFRRAKQ
MW:2182.5 Da
% peak area by HPLC:95
Storage condition:-20° C