Mouse MBP (4-14)
Catalog # 102999-378
Supplier: Anaspec Inc
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Species:Mouse
- Size:5 mg
- Storage conditions:–20 °C
- Protein synonyms:Myelin basic protein 4-14
- Protein/peptide name:MBP (4-14)
- Purity:95%
- Molecular weight:1390.6 Da
- Sequence:QKRPSQRSKYL
- Cat. no.:102999-378
- Supplier No.:AS-22916
Specifications
About this item
This sequence corresponds to amino acids 3-13 from murine MBP. MBP induces immune reactivity in multiple sclerosis (MS), a chronic inflammatory demyelinating disease of the CNS. MBP (3-13) is a substrate for Protein Kinase C.
Sequence:QKRPSQRSKYL
MW:1390.6 Da
% peak area by HPLC:95
Storage condition:-20° C