Order Entry
Canada
ContactUsLinkComponent
GRGDSPK, AnaSpec
GRGDSPK, AnaSpec
Catalog # 103006-348
Supplier:  Anaspec Inc
CAS Number:  
GRGDSPK, AnaSpec
Catalog # 103006-348
Supplier:  Anaspec Inc
Supplier Number:  AS-61109
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    GRGDSPK Peptide|RGD Peptide
  • Protein/peptide name:
    GRGDSPK
  • Purity:
    95%
  • Molecular weight:
    715.8 Da
  • Sequence:
    GRGDSPK
  • Cat. no.:
    103006-348
  • Supplier No.:
    AS-61109

Specifications

About this item

A linear integrin binding peptide. It inhibits endothelial cell adhesion to fibroblast growth factor-2 and to fibronectin.
Sequence:GRGDSPK
MW:715.8 Da
% peak area by HPLC:95
Storage condition:-20° C