Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Size:5 mg
- Storage conditions:–20 °C
- Protein synonyms:GRGDS Peptide|Osteopontin sequence|cell adhesion Peptide
- Protein/peptide name:GRGDS
- Purity:95%
- Molecular weight:490.5 Da
- Sequence:GRGDS
- Cat. no.:103006-344
- Supplier No.:AS-61107
GRGDS is the cell adhesion sequence of osteopontin that recognizes the avb3 integrin. Osteopontin is overexpressed in experimental models of malignancy.
Sequence:GRGDS
MW:490.5 Da
% peak area by HPLC:95
Storage condition:-20° C